Invitrogen™

FluoReporter™ lacZ Flow Cytometry Kit

Catalog number:  F1930

 Related applications: Cellular Imaging | Flow Cytometry | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The FluoReporter® lacZ Flow Cytometry Kit provides materials and protocols for quantitating β-galactosidase activity with fluorescein di-V-galactoside (FDG) in single cells using flow cytometry. This kit is accompanied by a license to practice patented techniques for loading FDG by hypotonic shock and improving retention of fluorescein in lacZ-positive cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: β-Gal (lacZ) Assay
Detection Method: Fluorescent
Excitation⁄Emission (nm): 491⁄514
For Use With (Equipment): Flow Cytometer
Form: Solution
Format: Tube(s)
Number of Reactions: 50
Product Line: FluoReporter®
Product Size: 1 kit
Substrate: FDG
Substrate Properties: Chemical Substrate
Substrate Type: Beta-Gal Substrate
Target Enzyme: Beta-Galactosidase
Shipping Condition: Room Temperature

Contents & storage

Contains 1 vial FDG (0.5 mL, 20 mM in water/DMSO/ethanol, 8:1:1, v/v), 1 vial PETG (2.0 mL, 50 mM in water), chloroquine diphosphates (1.0 mL, 30 mM in water) and 1 vial of propidium iodide (1.0 mL, 150 µM in water).

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products