Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

5(6)-SFX (6-(Fluorescein-5-(and-6)-Carboxamido) Hexanoic Acid, Succinimidyl Ester), mixed isomers

Numéro de catalogue:  F2181

Invitrogen™  Applications liées: Labeling Chemistry | Protein Labeling & Crosslinking

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

Fluorescein is a common green-fluorescent derivatization reagent, while succinimidyl esters are frequently used for the labeling of the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. This "SFX" succinimidyl ester contains a seven-atom amino-hexanoyl spacer between the fluorophore and the reactive group. This spacer separates the fluorophore from the biomolecule to which it is conjugated, potentially reducing the quenching that typically occurs upon conjugation. The resulting fluorescein conjugates can be detected with standard FITC or GFP filter sets. Fluorescein-based dyes and their conjugates have a relatively high rate of photobleaching and a pH-sensitive fluorescence. Alexa Fluor® 488 and Oregon Green® 488 are alternative dyes whose spectra mimic those of fluorescein and are much more photostable with little to no pH sensitivity in the physicological pH range.

Specifications:

• Fluorophore label : fluorescein

• Reactive group: succinimidyl ester

• Reactivity: primary amines on proteins and ligands, amine-modified oligonucleotides

• Molecular weight: 586

• Extinction coefficient: 74,000 cm-1M-1

• Ex/Em of the conjugate: 494/520 nm

• Spectrally similar dyes: Alexa Fluor® 488, GFP

Typical Conjugation Reaction

Amine-reactive reagents can be conjugated with virtually any protein or peptide; the provided protocol is optimized for IgG antibodies. You may scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The fluorescein succinimidyl ester is typically dissolved in high-quality, anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) and the reaction carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification

Labeled antibodies are typically separated from free dye using a gel filtration column, such as Sephadex™ G-25, BioGel® P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis.

Alternative Packagings

Mixed-isomer amine-reactive succinimidyl esters of fluorescein are also available in a 10 x 1 mg unit size (F6129), a single isomer (F6106), and with a spacer that is more hydrophilic than the SFX spacer (F6130), as well as in numerous kits for labeling proteins and nucleic acids (Active esters and kits for labeling proteins and nucleic acids—Table 1.2).
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Label or Dye: FITC (Fluorescein)
Chemical Reactivity: Amine
Product Size: 10 mg
Shipping Condition: Room Temperature

Contenu et stockage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuels et protocoles

Produits recommandés