Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
Invitrogen™

FluoZin™-1, Tripotassium Salt, cell impermeant

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The FluoZin™-1 and FluoZin™-2 indicators are designed to detect Zn2+ concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists. Based on the N-(2-methoxyphenyl)iminodiacetate chelator, these indicators are designed for detection of Zn2+ in the 0-100 µM range with minimal interfering Ca2+ sensitivity. FluoZin™ indicators are offered in cell-permeant and cell-impermeant forms.

Fluorescent Zinc Indicators Specifications:
• Label (Ex/Em): FluoZin™-1 (~495/515 nm)
• Kd (Zn2+) (in buffer): ~8 µM
• Lyophilized product may be dissolved in distilled water or aqueous buffer for use
• Product is typically loaded into cells using microinjection or diffusion from patch pipette


Find Fluorescent Indicators for Zinc and Other Metal Ions
We offer a number of fluorescent indicators to determine polyvalent cation concentrations inside cells, to follow metal ion transport through ion channels, to make measurements in environmental samples, and more. Review Fluorescent Indicators for Zn2+ and Other Metal Ions-Section 19.7 in the Molecular Probes® Handbook for more information on these products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Zinc Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope
Cell Permeability: Cell-Impermeant
Detection Method: Fluorescent
Product Line: FluoZin™
Product Size: 500 µg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols