BioProbes™ Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!
Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Invitrogen™

FISH Tag™ DNA Green Kit, with Alexa Fluor™ 488 dye

Catalog number:  F32947

 Related applications: Cellular Imaging | Labeling Chemistry | Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

FISH Tag™ DNA Kits provide a complete workflow solution for fluorescence in situ hybridization (FISH) applications. Each kit provides all of the reagents needed for probe synthesis, labeling, and purification, as well as for imaging the labeled specimen. Choose from FISH Tag™ DNA Kits in three Alexa Fluor® colors or get the FISH Tag™ DNA Multicolor Kit, which contains four spectrally distinct dyes to allow simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and for in situ analysis of mitochondrial DNA localization and content.

FISH Tag™ Kit Specifications:
• Dye (Ex/Em): Alexa Fluor® 488 (492/520 nm)
• Comprehensive kit includes 16 reagents
• Comes with detailed protocol to help ensure your success


Complete Workflow Solution for FISH
FISH Tag™ Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor® dyes. PureLink® nucleic acid purification technology is used for fast and efficient purification of the labeled probe, and SlowFade® Gold antifade reagent is added to preserve the fluorescent signal during imaging.

More Options for Nucleic Acid Labeling
To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes® Handbook.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
Detection Method: Fluorescent
Label or Dye: Alexa Fluor™ 488
Product Line: Alexa Fluor™, FISH Tag™
Product Size: 1 kit
Target Molecule: DNA
Target Specificity: Not Target-Specific
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products