Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

FocalCheck™ Fluorescence Microscope Test Slide #1, for alignment, intensity, & calibration

Catalog number:  F36909

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The FocalCheck™ fluorescence microscope test slide #1 is specifically designed for calibrating fluorescence microscope systems and evaluating system and filter performance. The slide contains 10 sample areas coated with FocalCheck™ ring stained microspheres ranging in size from 0.5-15 µm that provide a visual representation of instrument misalignment or other aberrations in row A; row B contains a series of microspheres with fluorescence intensities ranging from 0.667-100%.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibration Type: Confocal Microscope Calibration, Fluorescence Microscope Calibration
Color: Assorted Colors
Format: Suspension Beads
Product Line: FocalCheck™
Product Size: 1 each
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products