Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.

FocalCheck™ Fluorescence Microscope Test Slide #3, 5 colors, high & low intensities

Catalog number:  F36914

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The FocalCheck™ fluorescence microscope test slide #3 is specifically designed for calibrating fluorescence microscope systems and evaluating system and filter performance. This slide is useful for basic evaluation of filter performance and as a general practice slide for fluorescence microscopy and digital imaging.
For Research Use Only. Not for use in diagnostic procedures.


Calibration Type: Confocal Microscope Calibration, Fluorescence Microscope Calibration
Color: Assorted Colors
Format: Suspension Beads
Product Line: FocalCheck™
Product Size: 1 each
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.


Manuals & protocols

Recommended products