Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

NBD Fluoride; 4-Fluoro-7-Nitrobenzofurazan, 4-Fluoro-7-Nitrobenz-2-Oxa-1,3-Diazole

Catalog number:  F486

Invitrogen™  Related applications: Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NBD fluoride can be used to attach the environment sensitive fluorophore, NBD, to primary amines, however, it can also react with thiols and cysteine to yield thioethers. NBD fluoride can also react with secondary amines. NBD is a functional analog of the dinitrophenyl hapten, and its fluorescence is quenched upon binding to anti-dinitrophenyl (anti-DNP) antibodies.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: NBD
Chemical Reactivity: Amine
Product Size: 25 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products