Fura-2 Calcium Imaging Calibration Kit (Zero to 10 mM CaEGTA, 50 µM Fura-2)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Fura-2 Calcium Imaging Calibration Kit is designed to facilitate rapid calibration and standardization of digital imaging microscopes. The kit contains 11 pre-diluted buffers supplemented with 50 µM fura-2 (F-1200) as well as 15 µm—diameter microspheres.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Ion Calibration Reagent
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope
Detection Method: Fluorescent
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products