Invitrogen™

IC Fixation Buffer

Catalog number:  FB001

 Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Intracellular fixation buffer for use in flow cytomerty, particularly for fixation in intracellular staining procedures. Composition is paraformaldehyde in phosphate buffered saline, pH 7.3. These Make-to-Order catalog items have a 2 week lead time.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Compatible Products: cell surface antibodies and intracellular cytokine antibodies
Product Size: 100 mL
Reagent Type: Fixation Reagent
Volume (Metric): 100 ml
Shipping Condition: Wet Ice

Contents & storage

100 mL bottle of IC Fixation Buffer.
Store reagents at 2-8°C. Do not freeze.

Documents

Manuals & protocols

Recommended products