TransFix® Cellular Antigen Stabilizing Reagent

Catalog number: FIX2

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Introducing TransFix® Reagent - a whole blood preservative that stabilizes leukocytes and preserves antigenic sites for future cellular analysis (Figure 1). Now it's simple to batch-process multiple samples in order to optimize laboratory workflow. Additionally, blood that has been stabilized with TransFix® Reagent can be collected and transported to other sites for analysis. TransFix® Reagent is:

• Easy to use-just add to whole blood, mix, and store for up to 10 days at 4°C
• Cost effective-batch samples to optimize laboratory work-flow while maintaining sample integrity
• Proven-multiple publications demonstrate its effectiveness at preserving a broad range of cellular antigens (Table 1)
• Convenient-compatible with EDTA (K3), ACD, and heparin anticoagulants
• Flexible-preserves a wide range of samples, including human, mouse, rat, sheep, pig, and more

View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads® for cell isolation.

References
1. Barnett, D. et al. (1999) Cytometry 38:88;
2. Jani, I.V. et al. (2001) J Immunol Methods 257:145;
3. Bergeron, M. et al. (2002) Cytometry 50:86.

TransFix® Reagent is a proprietary whole blood stabilization solution, and is a general purpose reagent (GPR).
TransFix® is a registered trademark of Sheffield Teaching Hospitals NHS Trust.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: TransFix®
Product Size: 2 mL
Reagent Type: Fixation Reagent
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products