Gelatin From Pig Skin, Fluorescein Conjugate

Catalog number:  G13187

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Molecular Probes® fluorescent conjugates of gelatin are designed for researchers studying not only collagen-binding proteins and collagen metabolism, but also for the study of gelatinases and collagenases (metalloproteins that digest collagen and gelatin). We offer two fluorescent conjugates of gelatin—one in which gelatin is coupled to fluorescein (G13187) and the other in which gelatin is coupled to our Oregon Green® 488 dye (G13186).

Gelatin Conjugate Specifications:
• Label (Ex/Em): Fluorescein (~499/519 nm)
• Degree of labeling is listed on the product label (based on molecular weight of 100,000)


Find More Probes for Following Receptor Binding and Cell Adhesion
Review Probes for Following Receptor Binding and Phagocytosis—Section 16.1 and Probes for Cell Adhesion, Chemotaxis, Multidrug Resistance and Glutathione—Section 15.6 in the Molecular Probes® Handbook for more information on these products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 5 mg
Protein Subtype: Gelatin
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols