Guanosine 5'-O-(3-Thiotriphosphate), BODIPY™ FL Thioester, Sodium Salt (BODIPY™ FL GTP-γ-S, Thioester)

Catalog number: G22183

Invitrogen™  Related applications: Pharma & Biopharma | Protein Assays and Analysis | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

BODIPY FL GTP-γ-S binds to GTP-binding proteins. To create this fluorescent nucleotide, the BODIPY FL fluorophore was linked through the γ-thiol of GTP-γ-S. The fluorescence of BODIPY FL GTP-γ-S is quenched ~90% relative to that of the free dye, but may be recovered on binding to G-proteins. In addition to its potential use for binding studies, the BODIPY FL GTP-γ-S thioether is an important new substrate for Fhit, a member of the histidine triad superfamily of nucleotide-binding proteins.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: 5 mM
Label or Dye: BODIPY® FL
Labeling Method: Chemical Labeling, Direct Labeling
Product Line: BODIPY™
Product Size: 50 µL
Shipping Condition: Wet Ice

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols