Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.

Glutathione Agarose, Linked Through Sulfur, sedimented bead suspension

Catalog number: G2879

Molecular Probes™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Glutathione agarose prepared by linking the sulfur atom of glutathione to crosslinked beaded agarose can be used for purification of GST fusion proteins. Each milliliter of gel can bind approximately 5-6 mg of boivine-liver GST. Adding excess free glutathione liberates the GST fragment from the matrix which can then be regenerated by washing with a high-salt buffer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: Suspension
Column Type: Affinity Column
Product Size: 10 mL
Immobilized Phase: Glutathione
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products