E-Gel® EX Agarose Gels, 1%

Catalog number: G402001

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

E-Gel® EX Gels are the fastest-resolving gels in the E-Gel® product line. They are pre-cast 1% and 2% gels in the E-Gel® format with 11 wells and a novel, openable format. The gels contain a proprietary fluorescent nucleic acid stain (excitation: 490 nm, emission: 520 nm) that can be viewed with blue light, which minimizes DNA damage and allows detection down to 1 ng/band of DNA.

Features of E-Gel® EX gels:

• Complete separation in just 10 minutes
• Ultra-sensitive detection of DNA
• Quick check of RNA samples
• Live monitoring of DNA migration without the hazards of UV light
• Easy access to the gel for extraction of DNA bands or transfer to membranes for Southern blotting

Ultra-sensitive detection of DNA
E-Gel® EX gels were developed for ultimate sensitivity and demonstrate over 5-fold greater sensitivity than comparable gels containing ethidium bromide. The superior sensitivity allows you to use lower amounts of sample, saving time and money.

Quick check of RNA samples
E-Gel® EX gels can also be used for fast analysis of RNA samples to check on their integrity before proceeding with downstream applications.

Live monitoring of DNA migration
A proprietary stain incorporated in the gels is excitable by visible blue light, allowing safe, real-time monitoring of DNA or RNA migration with the E-Gel® Safe Imager Real-Time Transilluminator.

Easy access to the gel
The gel cassette is designed to open with a Gel Knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis.

Nucleic acid markers
The room-temperature stable, ready-to-use E-Gel® 1 Kb Plus DNA Ladder is premixed with loading buffer and is formulated specifically for maximum performance on E-Gel® EX gels as well as other E-Gel® precast gels. Standard ladders not premixed with loading buffer may also be used, but in significantly lower amounts than required for other gels. 0.1–2 Kb and 0.5–10 Kb RNA ladders may be used when separating RNA samples.

Running and viewing apparatus
E-Gel® EX gels require the E-Gel® iBase™ Power System for running and the E-Gel® Safe Imager™ Real-Time Transilluminator for real-time viewing of sample migration.

E-Gel® EX gel formats
E-Gel® EX gels are offered in 10- and 20- pack sizes. Starter Kits are an economical way to get started and include gels, devices, and accessories.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Percentage: 1 %
Gel Type: E-Gel®
In-Gel Dye: Proprietary Stain
Mode of Separation: Molecular Weight
Product Line: E-Gel®
Product Size: 20 gels
Sample Buffer Type: Non-preparative, Preparative without dyes
Sample Loading Volume: 20 µL
Sample Type (Specific): Double-Stranded DNA (dsDNA), Single-Stranded RNA
Separation Range: 100 bp to 5 kb
Well Format: Single Comb
Wells: 11 wells
Shipping Condition: Room Temperature

Contents & storage

E-Gel® EX 20-Paks contain 20 gels. Store gels at room temperature. E-Gel® EX gels are guaranteed stable for 3 months when properly stored.

Documents

Manuals & protocols

Recommended products