E-Gel™ Double Comb Agarose Gels, 0.8%

Catalog number: G601808

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

E-Gel® Double Comb Gels allow you to resolve 16 samples in just 15 minutes. The E-Gel® double comb gel is supplied in a two-row, mini-gel format. Each row contains 8 sample wells and 1 well for a marker. The spacing between wells is compatible with eight-channel pipettors for easy, high-throughput loading. E-Gels are available in two pack sizes—a Starter Pak (6 E-Gels) or an 18-Pak (18 E-Gels). Agarose percentages (0.8%, 1.2%, and 2%) are general purpose gels and the 4% agarose option is used for high-resolution separation.

About E-Gel® agarose gels
E-Gels are self-contained, pre-cast agarose gels designed to provide fast, convenient, and easy electrophoresis. Each E-Gel® is ready for use with agarose, electrodes, and DNA stain (ethidium bromide or SYBR® Safe) packaged inside a dry, disposable, UV-transparent cassette. There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble. Just load your samples and run. E-Gels offer excellent resolution and clarity in as little as 12 minutes and are ideal for analyzing PCR products, restriction digests, and plasmid preparations. E-Gels are available in a variety of formats and gel percentages.

Specifications

Gel Percentage: 0.8 %
Gel Type: E-Gel®
In-Gel Dye: Ethidium Bromide
Product Line: E-Gel®
Product Size: 18 gels
Well Format: Double Comb
Wells: 2 x 8 wells
Shipping Condition: Room Temperature

Contents & storage

Each standard box of E-Gel® Double Comb Gels contains 18 gels and a manual. Store at room temperature.

Documents

Manuals & protocols