E-Gel® 96 Agarose Gels, 1%

Catalog number: G700801

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

E-Gel® 96 Agarose Gels are bufferless, pre-cast gels designed for fast, high-throughput DNA electrophoresis. Each gel contains 96 sample lanes and 8 marker lanes that are ideal for screening multiple PCR products, plasmid preparations, and restriction digests. The unique 96-well staggered-well format provides a 1.6 cm run length that resolves between 100 bp and 10 kb. The E-Gel® 96 loading format is compatible with multi-channel pipettors, and the most commonly used 8-, 12-, and 96-pin liquid handling robots (see figure). With 12-minute run times, up to 20,000 samples can be resolved in a single day.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Wells: 96 wells
Gel Type: E-Gel®
In-Gel Dye: Ethidium Bromide
Product Size: 8 gels
Gel Percentage: 1 %
Shipping Condition: Room Temperature

Contents & storage

Each box of E-Gel® 96 gels contains 8 gels and a manual. Store at room temperature.

Documents

Manuals & protocols

Recommended products