FIX & PERM® Cell Permeabilization Kit

Catalog number: GAS003

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The FIX & PERM® Cell Permeabilization Kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.

The FIX & PERM® Cell Permeabilization Kit is:

• Flexible—compatible with multiple sample types and cellular antigens
• Robust—scatter pattern of cells unaffected
• Efficient—simultaneous detection of intracellular and surface markers

View a selection guide for all available FIX & PERM® products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads® cell separation products for cell isolation).

The FIX & PERM® formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.The FIX & PERM® Cell Permeabilization Kit is suitable for various biological samples and has been validated for use with cell lines, whole blood, and bone marrow. The FIX & PERM® kit has been shown to accurately identify previously undetectable intracellular markers. Unlike other fixation and permeabilization reagents available on the market, the FIX & PERM® staining protocol leaves the morphological scatter characteristics of the cells intact.

How It Works
The FIX & PERM® Cell Permeabilization Kit contains 2 reagents: Fixation Medium (Medium A) and Permeabilization Medium (Medium B). Cells are incubated for 15 minutes at room temperature with Fixation Medium, washed, and then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Fixation Medium and Permeabilization Medium are also available separately (Cat. Nos. GAS001S100 & GAS002S100).

Specifications

Product Line: FIX & PERM®
Product Size: 50 tests
Reagent Type: Fixation Reagent, Permeabilization Reagent
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products