Invitrogen™

Lysing Solution, Whole Blood Lysing Solution

Catalog number:  GAS010

 Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Invitrogen Cal-Lyse™ is a lysing solution formulated to enable the lysis of erythrocytes in samples of anticoagulated human peripheral blood. Invitrogen Cal-Lyse™ lysing solution is intended to be used as an aid in the enumeration of leukocytes that have been stained with monoclonal antibodies for flow cytometric analysis.

View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads® for cell isolation.
For In Vitro Diagnostic Use.

Specifications

Reagent Type: Whole Blood Lysis Buffer
Product Size: 25 mL
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products