Trap-Pak™ Bags (Medium)

Catalog number: GEN084033

Applied Biosystems™  Related applications: Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to reduce and maintain a low water content in solutions that must remain anhydrous.

Used With:
Anhydrous Acetonitrile, Amidite Diluent, Tetrazole⁄Acetonitrile, Amidite Activator Solution, Tetrazole⁄Acetonitrile, Amidite Activator Solution, Tetrazole⁄Acetonitrile, Amidite Activator Solution

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 3900 DNA Synthesizer
Form: Solid
Product Line: Applied Biosystems™, Trap-Pak™
Product Size: 1 package
Shipping Condition: Room Temperature

Contents & storage

This product may be stored at room temperature.

Documents

Manuals & protocols