Gibco™

Rat MRP2 Vesicles

Catalog number:  GM0002

 Related applications: Pharma & Biopharma | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Rat MRP2 Vesicles are insect derived purified plasma membranes with inserted MRP2 transport protein (multidrug resistance-associated protein, ABCC2). Our Rat MRP2 Vesicles can be used to evaluate test compounds and drug interactions with MRP2 in in vitro assays. MRP2 is an ATP-binding cassette (ABC) transporter that carries organic anions.

Use Rat MRP2 Vesicles to:

• Investigate the transporter interactions of your drug candidates
• Assess potential for transporter-mediated drug-drug interactions
• Obtain high quality results with a large signal to noise ratios

Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate’s substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these "inside-out" vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

Transporter Vesicle Details
Rat MRP2 Vesicles are part of a larger group of transporter vesicles known as ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells transfected with baculovirus) expressing MRP2. MRP2 is expressed in various organs, and is thought to play a role in excretion of various compounds from the kidney and liver, based on its localization on the canalicular plasma membrane of hepatocytes and brush border membrane of tubular epithelial cells.

For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: Vial(s)
Form: Frozen
Product Size: 0.5 mL
Purity or Quality Grade: Research Grade
Shipping Condition: Dry Ice
Species: Rat

Contents & storage

Store at -80°C.

Documents

Manuals & protocols

Recommended products