Control Vesicles

Catalog number: GM0003

Gibco™  Related applications: Pharma & Biopharma | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These Control Vesicles are insect-derived purified plasma membranes with no inserted transport proteins for use as a negative control in vesicular transport assays. Our Control Vesicles control for nonspecific absorption and transport of test substrates to insect plasma membranes.

Control Vesicle Details
Control Vesicles are a negative control product for use with ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells). Due to the nonspecific transport activity and absorption properties of test substrates to Sf9 cell membranes use of these Control Vesicles is highly recommended.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Type: Negative Control
Format: Vial(s)
Form: Frozen
Product Size: 0.5 mL
Purity or Quality Grade: Research Grade
Shipping Condition: Dry Ice
Species: S. frugiperda, Spodoptera frugiperda

Contents & storage

Store Control Vesicles at -80°C.

Documents

Manuals & protocols

Recommended products