Human MDR1 Vesicles

Catalog number:  GM0015

Gibco™  Related applications: Pharma & Biopharma | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Human MDR1 Vesicles are insect derived purified plasma membranes with inserted MDR1 transport protein (P-glycoprotein, ABCB1). Our Human MDR1 Vesicles can be used to evaluate test compounds and drug interactions with MDR1 in in vitro assays. MDR1 is an ATP-binding cassette (ABC) transporter associated with multidrug resistance, including resistance to anti-cancer drugs.

Use Human MDR1 Vesicles to:

• Investigate the transporter interactions of your drug candidates
• Assess potential for transporter-mediated drug-drug interactions
• Obtain high quality results with a large signal to noise ratios

Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate's substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these "inside-out" vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

Transporter Vesicle Details
Human MDR1 Vesicles are part of a larger group of transporter vesicles known as ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells transfected with baculovirus) expressing MDR1. MDR1 is an important determinant of drug disposition, through its role in processes such as absorption at the small intestine and transfer across the blood-brain barrier to the CNS.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: Vial(s)
LoyaltyEligible: SKUEligibleForLoyalty
Form: Frozen
Product Size: 0.5 mL
Purity or Quality Grade: Research Grade
Shipping Condition: Dry Ice
Species: Human

Contents & storage

Store at -80°C.

Documents

Manuals & protocols

Recommended products