Gibco™

MRPs-BCRP Vesicular Transport Assay Reagent Set

Catalog number:  GM3010

 Related applications: Pharma & Biopharma | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The MRPs-BCRP Vesicular Transport Assay Reagent Set is a ready-to-use kit containing all reagents needed for vesicular transport assay except for the vesicles themselves. The reagent set can be used for vesicular transport assay using radioisotope-labeled or non-labeled compound. This kit has enough material to perform 100 vesicular transport assays.

Use The MRPs-BCRP Vesicular Transport Assay Reagent Set to:

• Investigate the transporter interactions of your drug candidates
• Assess potential for transporter-mediated drug-drug interactions
• Obtain high quality results with a large signal to noise ratios

Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate’s substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these "inside-out" vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

Kit Compatibility
Our MRPs-BCRP Vesicular Transport Assay Reagent Set is designed for use with either MRP or BCRP Vesicles. The 100 vesicular transport assays that can be performed with this kit will require two MRP or BCRP Vesicle products.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Frozen
Format: Bottle(s)
Product Size: 100 assays
Shipping Condition: Dry Ice

Contents & storage

Store at -20°C.

Documents

Manuals & protocols