Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

HCS LIVE/DEAD® Green Kit

Catalog number: H10290

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The HCS LIVE⁄DEAD® Green Kit measures cytotoxicity by high content analysis with the Image-iT® DEAD Green™ viability stain – a non-fluorescent and cell impermeant compound that exhibits a strong fluorescent enhancement upon binding to DNA. Drugs and test compounds leading to serious cell injuries, including plasma membrane permeability, allow entry of the Image-iT® DEAD Green™ viability stain. This two-color fluorescence assay also employs the use of a cell permeant nuclear segmentation tool that stains both live and dead cells, reflecting the total cell number within the sample. For total cell staining, the kit includes both a blue-fluorescent Hoechst 33342 and a near infrared-fluorescent, HCS NuclearMask™ Deep Red reagent. Either of these segmentation tools may be used with Image-iT® DEAD Green™ viability stain for excellent discrimination of live and dead cells that survives fixation and permeabilization for combination with additional parameters of cytotoxicity or other biomarkers. The kit contains sufficient reagents for performing 2, 96-well plates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Compatible Cells: Eukaryotic Cells
For Use With (Equipment): Fluorescence Microscope, High Content Instrument
Detection Method: Fluorescent
Format: 96-well plate
Color: Blue, Far-Red, Green
Emission Class: UV, Visible, near IR
Excitation Class: UV, Visible, near IR
Excitation⁄Emission (nm): 350⁄461, 488⁄515, 638⁄686
Label or Dye: HCS NuclearMask™ Deep Red stain, Hoechst 33342, Image-iT® DEAD Green viability stain
Product Line: LIVE/DEAD®, Molecular Probes®
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store at ≤-20°C, desiccate and protect from light.

Documents

Manuals & protocols

Recommended products