Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

DiIC1(5) (1,1',3,3,3',3'-Hexamethylindodicarbocyanine Iodide)

Catalog number:  H14700

Invitrogen™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The cationic carbocyanine DiIC1(5) can be used as a slow-response potential-sensitive probe. This dye accumulates on hyperpolarized membranes and is translocated into the lipid bilyaer. Aggregation within the confined membrane interior usually results in decreased fluorescence and absorption shifts. This far red-fluorescent dye exhibits excitation/emission maxima of ~638/658 nm.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Carbocyanines
Product Size: 100 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols