Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Hydroxystilbamidine, methanesulfonate

Catalog number: H22845

Invitrogen™  Related applications: Cell Tracing & Tracking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Hydroxystilbamidine exhibits distinctively different fluorescence emission profiles when bound to DNA compared to RNA. This cationic dye is also frequently used as a retrograde neuronal tracer. Please note that the product offered prior to July 2001 under catalog number H-7599 was actually aminostilbamidine and not hydroxystilbamidine. Aminostilbamidine is now offered under a new catalog number, A-22850.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Blue
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Nucleic Acids, Nucleus
Detection Method: Fluorescent
Excitation⁄Emission (nm): 385⁄variable with nucleic acid
Product Size: 10 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products