Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

HCS CellMask™ Deep Red Stain

Catalog number: H32721

Invitrogen™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

HCS CellMask™ Deep Red stain (H32721) is a cell delineation tool for HCS platforms. HCS CellMask™ stains label the entire cell and provide an accurate backdrop against which the features of interest can be assessed. HCS CellMask™ stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask™ stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask™ stains are also available in a choice of colors for multiplexing flexibility.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Deep Red, Far-Red
For Use With (Equipment): High Content Instrument
Sub-Cellular Localization: Cytoplasm & Cytosol, Nucleus
Detection Method: Fluorescent
Emission Class: near IR
Excitation Class: near IR
Excitation⁄Emission (nm): 650⁄655
Flow Cytometer Laser Lines: NA
Product Line: Molecular Probes®
Product Size: 1 set
Shipping Condition: Room Temperature

Contents & storage

1 set of vials, providing sufficient material to stain a total of 10, 96-well microplates.



Sufficient reagent is supplied to perform 10, 96-well plates. Store at ≤-20°C, desiccate and protect from light.

Documents

Manuals & protocols

Recommended products