Human Suspension Hepatocytes, Metabolism Qualified, Male, (4-8 x 106 cells)

Catalog number: HMCS1S

Gibco™  Related applications: Hepatocytes & Hepatic Models | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These cryopreserved human male hepatocytes are suitable for use in suspension metabolism applications.

Cells per vial: 4-8 million

The commonly tested Phase I (P450) and Phase II (UGT, SULT) drug metabolizing enzymes were analyzed for metabolic activity in each lot. Metabolite formation was measured by standard biochemical assays using LC⁄MS⁄MS or HPLC.

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
• Phase I and II enzyme activities
• Proper morphology
• Viabilities generally >75%

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links:

Learn more about our cryopreserved hepatocytes

Learn about our full line of hepatocytes ,

Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Cryopreserved
Species: Human
Quantity: 1 vial
Cell Type: Hepatocytes
Donor Source: Single Donor
Product Size: 1 vial
Number of Cells: 4-8 x 106

Contents & storage

Vials of cryopreserved hepatocytes should be stored in a cryostorage dewar or -135°C freezer.

Documents

Manuals & protocols