HepaRG™ Tox Medium Supplement

Catalog number: HPRG630

Gibco™  Related applications: Hepatocytes & Hepatic Models | Pharma & Biopharma | Primary Cell Culture | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

HepaRG™ Tox Medium Supplement is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Tox Medium Supplement is specifically used for applications such as short and long term toxicity studies that require maintenance of hepatocyte-specific functions. The HepaRG™ Tox Medium Supplement is specifically designed for use with HepaRG™ cells.
• Easy media preparation, just thaw, mix, and use
• Reproducible, and biologically relevant results with HepaRG™ cells

Media Preparation
Working media is prepared by the addition of thawed HepaRG™ Tox Medium Supplement to 100 ml of William’s Medium E (A1217601 or 12551032) and 1 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.

Simplified, Relevant and Reproducible Studies
HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

HepaRG™ is a trademark of BioPredic International.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: HepaRG™ Cells
Form: Frozen
Glutamine: No Glutamine
Product Line: Gibco®
Product Size: 1 bottle
Purity or Quality Grade: Cell Culture Grade, Research Grade
Serum Level: Standard Serum
Sterility: Sterile
Tests Performed: Sterility Testing (Bacteria & Fungi)
Volume (Metric): 14 ml
Shipping Condition: Dry Ice

Contents & storage

Supplement should be stored at -20°C

Documents

Manuals & protocols