Invitrogen™

Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 568 Conjugate

Catalog number: I21412
Invitrogen™

Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 568 Conjugate

Catalog number: I21412
Catalog Number
I21412
Unit Size
500 µg
Price (USD)
Price: 516.00
Online price: 490.65
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
I21412500 µg
Price: 516.00
Online price: 490.65
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4) isolated from the seeds of the tropical African legume Griffonia simplicifolia. The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The red fluorescent Alexa Fluor™ 568 isolectin GS-IB4 conjugate can be used in fluorescence microscopy.

View complete list of fluorescent dye-conjugated lectins ›
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
500 μg
Source
Africa Shrub legume
Conjugate
Alexa Fluor 568
Label Type
Alexa Fluor Dyes
Shipping Condition
Room Temperature
Form
Lyophilized
Protein Subtype
Lectins
Product Line
Alexa Fluor™

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text