Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
Invitrogen™
Ionomycin, Calcium Salt
Catalog number:  I24222
 Related applications: Cell Viability, Proliferation & Function
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Ionomycin is an effective Ca2+ ionopohre that is commonly used to both modify intracellular Ca2+ concentrations and to calibrate fluorescent Ca2+ indicators when studying the regulatory properties of Ca2+ in cellular processes. Ionomycin also transports Pb2+ and some other divalent cations, as well as several lanthanide series trivalent cations, at efficiencies that are greater than or equal to those for Ca2+.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Indicator Type: Ionophore
Product Size: 1 mg
Shipping Condition: Room Temperature
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols
Recommended products