Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.

ImaGene Green™ C12FDGlcU GUS Gene Expression Kit

Catalog number: I2908

Invitrogen™  Related applications: Cellular Imaging | Flow Cytometry | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ImaGene Green™ GUS Gene Expression Kit contains a fluorescein-based glucuronidase substrate that has been covalently modified to include a 12-carbon lipophilic moiety, C12FDGlcU. Once inside the cell, the substrates are cleaved by ß-glucoronidase, producing a fluorescent product that is well retained by the cells, probably by incorporation of the lipophilic tail within the cellular membrane. In addition to the C12FDGlcU, the kits also include the broad-spectrum ß-glucoronidase inhibitor, D-glucaric acid-1,4-lactone.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: GUS (β-Glucuronidase) Assay
Excitation⁄Emission (nm): 495⁄518
Product Line: ImaGene Green™
Product Size: 1 kit
Substrate Properties: Chemical Substrate
Substrate Type: Beta-Glucuronidase (GUS) Substrate
Target Enzyme: Beta Glucuronidase (GUS)
Technique: Fluorescence Intensity
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products