Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Image-IT® LIVE Mitochondrial and Nuclear Labeling Kit (Counterstains For GFP-Expressing Cells)

Catalog number: I34154

Molecular Probes™  Related applications: Cell Viability, Proliferation & Function | Cellular Imaging | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Image-iT™ LIVE Mitochondrial and Nuclear Labeling Kit provides two stains - red-fluorescent MitoTracker® Red CMXRos dye and blue-fluorescent Hoechst 33342 dye for highly selective mitochondrial and nuclear staining, respectively in live green-fluorescent protein (GFP)-transfected cells. Using the protocol provided, the dyes can be combined into one staining solution to save labeling time and wash steps while still providing optimal staining.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Blue, Red
Detection Method: Fluorescent
For Use With (Equipment): Fluorescence Microscope
Product Line: Image-iT®
Product Size: 1 kit
Sub-Cellular Localization: Mitochondria, Nucleus
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products