Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Image-iT™ FX Signal Enhancer

Catalog number: I36933

Invitrogen™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Image-iT® FX signal enhancer is a liquid that is applied directly to slides or coverslips containing fixed and permeabilized cell or tissue samples prior to staining with fluorescent probes. When Image-iT® FX signal enhancer is used, nonspecific fluorescence (background) commonly seen with the application of fluorescent conjugates of streptavidin, goat anti-mouse, or goat anti-rabbit IgG is largely eliminated.

Key Attributes

Easy protocol—apply enhancer from ready-made solution in dropper bottle
Effective with many fluorescent labels [see Table 2 in the product manual]
Cost effective—supplies enough for 50 standard coverslips
Compatible with your immunostaining protocol’s other blocking steps

Reduce Nonspecific Binding of Dyes to Tissues.
Alexa Fluor® dyes and many other dyes have negatively charged modifications that can lead to nonspecific association with cells and tissue. While not always significant, it can be a problem when pushing the limits of sensitivity in low-abundance targets. Image-iT® FX signal enhancer can significantly reduce this binding, resulting in marked improvements in staining.

Need help deciding what product you need for your cell imaging? See our cell imaging workflow and decision tree.

Tired of poor signal-to-noise? See our collection antifade and signal enhancer products from Molecular Probes®.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Signal Enhancer
Product Line: Image-iT®
Product Size: 10 mL
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products