Cat. No. IB701001 is a replacement for Cat. No. IB601001

iBlot® Western Detection Stacks, regular size

Catalog number: IB701001

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The iBlot® Western Detection Kit uses the iBlot® Dry Blotting System to speed up your western detection. Reduce the time needed for Western detection from overnight to typically less than 30 minutes. The iBlot® Western Detection Stacks comes with only the iBlot® anode and cathode stacks specific for western detection.
 Number of stacks: 10
•  Size: Regular (13.5 cm x 8 cm)

Note: This is not a complete kit. The iBlot® Western Detection Stacks come with only the iBlot® anode and cathode stacks. No western detection reagents are provided.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Related Links
•  Learn more about iBlot® Western Detection Stacks.
•  Learn more about the iBlot® Dry Blotting System.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

System: iBlot®
Gel Size: Midi (8 cm x 13 cm)
Membrane: Membrane Not Included
Product Size: 1 box
Gel Compatibility: E-PAGE™, Novex® Midi Gels
Shipping Condition: Room Temperature

Contents & storage

1 box: Stack Box to be kept at Room Temperature (5-25 degrees Celsius)

Documents

Manuals & protocols

Recommended products