Cat. No. IB731001 is a replacement for Cat. Nos. IB631003 and IB631001.

iBlot® Western Detection Chromogenic Kit, anti-mouse, regular

Catalog number: IB731001

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The iBlot® Western Detection Kit uses the iBlot® Dry Blotting System to speed up your western detection. Reduce the time needed for western detection from overnight to typically less than 30 minutes. The iBlot® Western Detection Kit comes with iBlot® anode and cathode stacks specific for western detection and the reagents for visualizing your proteins. Pick the size, secondary antibody, and detection method appropriate for your research.
 Size: Regular (13.5 cm x 8 cm)
•  Secondary antibody: Anti-Mouse
•  Detection Method: Chromogenic

Antibody Matrix
The iBlot® Western Detection antibody matrix (Fig 1) is used to block your membrane, and transfer primary and secondary antibodies to the membrane. Blocking takes two minutes and each transfer step takes three minutes (see Protocol).

Note: A fresh antibody matrix is used for each transfer step.

Visualize Your Protein by Chromogen or Chemiluminescence
After binding the primary and secondary antibodies to the membrane, use the included detection reagents for visualizing your antigens. You have your choice of chemiluminescent or chromogenic detection. The appropriate reagents for visualizing the antigen by chemiluminescent or chromogic detection are included.

The iBlot® Western Detection Kit is compatible with either PVDF or nitrocellulose membranes. The iBlot® Western Detection Kit is compatible with either membranes prepared by wet, semi-wet, dry, or iBlot® transfer.

Each kit comes with iBlot® Western Detection stacks and reagents for 10 membranes.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Related Links
•  Learn more about iBlot® Western Detection Stacks.
•  Learn more about the iBlot® Dry Blotting System.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Blue
Reactivity: Mouse
Label or Dye: AP (Alkaline Phosphatase)
Product Size: 10 pack
Substrate Type: AP (Alkaline Phosphatase) Substrate
Target Molecule: Protein
Detection Method: Chromogenic
Membrane Compatibility: PVDF, Nitrocellulose
Sample Type (Specific): Protein

Contents & storage

2 boxes: Reagent Box to be kept at 4 degrees Celsius; Stack Box to be kept at Room Temperature (5-25 degrees Celsius)

Documents

Manuals & protocols