iBlot™ USB Cable

Catalog number:  IB800105

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The iBlot® USB Cable enables uploading of new pre-set programs from the web site onto the iBlot® Gel Transfer Device
For Research Use Only. Not for use in diagnostic procedures.


For Use With (Equipment): iBlot™
Product Line: iBlot™
Product Size: 1 cable
Type: Cords and Connectors
Shipping Condition: Room Temperature

Contents & storage

1 USB cable


Manuals & protocols