Regulatory requirements vary by country; products may not be available in your geographic area.

LSIVet™ Bovine IBR Screening Serum ELISA Kit

Catalog number: IBRS10

 Related applications: Animal Health Immunoassays

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Same as IBRS/10

Single-well Indirect ELISA kits enable sensitive and specific detection of pathogen-specific antibodies in animal samples, allowing for reliable and rapid screening and detection of seropositive animals.

How the kit works:

1. Samples are added to the specific antigen-coated microplate. If present in the sample, specific anti-pathogen antibodies bind to the antigen and form antigen-antibody complexes.
2. After washing, an anti-species peroxidase-labeled conjugate is added, which binds to any antibodies attached to the plate.
3. After a second washing, a chromogenic substrate is added. Color development results from oxidation of the substrate by the conjugated peroxidase.
4. After stopping the reaction, results are read using an ELISA plate reader. Positive samples will show color. The amount of color in each well is proportional to the amount of specific antibodies in the sample.

For veterinary use only.

Regulatory requirements vary by country; products may not be available in your geographic area.

Biological hazard safety

WARNING! BIOHAZARD. Biological samples such as tissues, body fluids, infectious agents, and blood of humans and other animals have the potential to transmit infectious diseases. All work should be conducted in properly equipped facilities using the appropriate safety equipment (for example, physical containment devices). Safety equipment also may include items for personal protection, such as gloves, coats, gowns, shoe covers, boots, respirators, face shields, safety glasses, or goggles. Individuals should be trained according to applicable regulatory and company/institution requirements before working with potentially biohazardous materials. Follow all applicable local, state/provincial, and/or national regulations. The following references provide general guidelines when handling biological samples in laboratory environment.

U.S. Department of Health and Human Services, Biosafety in Microbiological and Biomedical Laboratories (BMBL), 5th Edition, HHS Publication No. (CDC) 21-1112, Revised December 2009

World Health Organization, Laboratory Biosafety Manual, 3rd Edition, WHO/CDS/CSR/LYO/2004.11
For Veterinary Use Only.

Specifications

Target Organism Class: Bovine Herpesvirus 1 (BHV-1)
Validated Application: ELISA
Detection Method: Colorimetric
Sample Type (Specific): Serum
Species: Cattle⁄Bovine
Product Size: 10 x 96 well plates

Documents

Manuals & protocols