Thermo Scientific™
Delta Ray™ CO2 Isotope Ratio Infrared Spectrometer with URI Connect
 Related applications: Industrial Mass Spectrometry
Thermo Scientific™ Delta Ray Connect™ IRIS with URI Connect represents a new and exciting solution that extends the continuous measurement of isotope ratios and concentrations of CO2 in air to discrete samples such as head space analysis or direct injection of small amounts of CO2. Utilizing state of the art mid-infrared spectroscopy, the simultaneous determination of δ13C and δ18O empowers scientists to make profound scientific discoveries in a wide variety of research fields requiring analysis of discrete samples such as water/aquifer research, paleoclimate research and microbiology.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Thermo Scientific™ Delta Ray Connect™ IRIS with URI Connect represents a new and exciting solution that extends the continuous measurement of isotope ratios and concentrations of CO2 in air to discrete samples such as head space analysis or direct injection of small amounts of CO2. Utilizing state of the art mid-infrared spectroscopy, the simultaneous determination of δ13C and δ18O empowers scientists to make profound scientific discoveries in a wide variety of research fields requiring analysis of discrete samples such as water/aquifer research, paleoclimate research and microbiology.
Get out more
Get out more
Push the limits of carbonate measurements
Learn more ›
Phenotyping With the Delta Ray IRIS
Documents
Manuals & protocols