iPrep™ ChargeSwitch® Forensic Kit This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.
Catalog number: IS10002
Invitrogen™  Related applications: Automated Nucleic Acid Purification | DNA Extraction | Forensics
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
• Flexibility in starting sample type, for quick and easy processing of different samples simultaneously
• High sensitivity with samples containing very low amounts of DNA
• Fast automated extraction time of 20 minutes
• Standardized procedures minimize human error; preset protocols and prefilled cartridges eliminate variation
The iPrep ChargeSwitch® Forensic Kit offers a single protocol for efficiently isolating DNA from all kinds of forensic samples (Table 1), even if
the DNA copy number is low or the DNA is partially degraded.
Designed specifically for use with the iPrep™ Purification Instrument, the iPrep™ ChargeSwitch® Forensic Kit takes advantage of the powerful ChargeSwitch® technology, a major advance in DNA purification. This technology is based on a unique, ionizable nucleic acid-binding ligand whose charge can be switched based on the pH of the surrounding medium (Figure 1). Purification is achieved through a simple three-step procedure in aqueous buffers, and avoids the use of guanidine, ethanol, and other troublesome reagents.
Contents and Storage:
The iPrep™ ChargeSwitch® Forensic Kit provides all reagents (supplied in prefilled cartridges), tubes, and tips needed for isolating DNA from forensic cell samples. Reagent cartridges can be stored at room temperature; store lysis buffer and proteinase K at 4°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Sample Type (General): Forensic Samples
Isolation Technology: ChargeSwitch® Chemistry
Downstream Application: Cloning, Forensic Testing, PCR, Sequencing, Southern Blotting
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Green Features: Fewer resources used, Less hazardous, Less waste
Product Line: ChargeSwitch®, iPrep™
Product Size: 52 preps
Documents
Manuals & protocols