iPrep™ PureLink® Total RNA Kit

Catalog number: IS10006

Ambion™  Related applications: Automated Nucleic Acid Purification | RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Choose the iPrep™ PureLink® Total RNA Kit for high yields from even the smallest sample (10 mg of tissue, 10e6 cultured cells or 1 ml of blood). The kit contains pre-filled, fully qualified cartridges to avoid any mix-ups or cross-contamination. It also provides low CVs and reduced purification variability, increasing the reliability of your gene expression assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Bacteria, Blood, Cells, Tissue, Yeast
High Throughput Compatibility: High Throughput-Compatible
Downstream Application: Microarray Analysis, Northern Blotting, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction
Final Product: Total RNA
Isolation Technology: Magnetic Bead
Product Line: PureLink™, iPrep™
Product Size: 1 kit
Shipping Condition: Room Temperature

Documents

Manuals & protocols

Recommended products