DiSBAC4(3)

Catalog number: K1022

 Related applications: Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Voltage Sensor Probes (VSPs) are a Fluorescence Resonance Energy Transfer (FRET)-based voltage-sensing assay technology used to measure changes in cellular membrane electrical potential. VSPs are ideal for screening compounds that modulate ion channels because they provide:

• High-throughput capabilities-screen 32,000 samples in one day
• Highly sensitive detection-accurately and precisely report a 1% change in the Emission Ratio per mV
• Rapid detection response-sub-second time response
• Ratiometric readout (the Emission Ratio)-reduces experimental errors arising from well-to-well variations in
• cell number, addition artifacts, dye loading, plate inconsistencies, temperature fluctuations, and signal intensities. Allows for highly accurate, real-time optical readout of changes in membrane potential
• Universal methodology-applicable to any target that changes the membrane potential
• Highly accurate, real-time optical readout-results correlate with patch clamp data

How VSPs Work
VSPs use a FRET pair composed of one of two highly fluorescent, mobile, voltage-sensitive acceptor oxonols [DiSBAC2(3) or DiSBAC4(3)] and a fluorescent, membrane-bound coumarinphospholipid FRET donor (CC2-DMPE). When the cell interior has a relatively negative potential, the oxonol dye will bind to the exterior of the cell membrane, resulting in efficient FRET. When the cell interior has a relatively positive potential, however,
the oxonol dye will bind to the interior of the cell membrane, thus separating the FRET pair and disrupting FRET.

DiSBAC2(3) is used for most ion channel screening applications because it is more sensitive, easier to load, more stable than DiSBAC4(3), and can remain in the extracellular media. The time response of this oxonol is ~500 ms. DiSBAC4(3) requires a surfactant for cellular loading and a washing step to remove excess dye, but responds to changes to the membrane potential in ~20 ms. This dye is useful for applications that require faster response time or no added dye in the extracellular solution.

VABSC-1 Dye (Voltage Assay Background Suppression Compound) performs in conjunction with CC2-DMPE to suppress background signal, providing optimal performance and minimal toxicity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 5 mg
Indicator Type: Voltage Sensor Probes
Indicator Properties: Fluorescent Dye-Based
Shipping Condition: Room Temperature

Contents & storage

The probe sets provide 1 mg of CC2-DMPE and 1 mg of either DiSBAC2(3) or DiSBAC4(3), enough reagents for approximately 20 96-well plates of assays when used according to the recommendations in the Product Guide on the Invitrogen website. Store at -20°C. VABSC-1 is provided as a 1 g size, which provides enough suppressor compound for about 100 plates. Store at room temperature. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols