T-REx™ Complete Kit, with pcDNA™4/TO/myc-His© Vector

Catalog number: K103001

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A Tetracycline-Regulated Expression System without Viral Transactivators
The T-REx™ System yields higher levels of induced expression than any other regulated mammalian expression system. It utilizes the complete CMV promoter and adds control elements from the bacterial tetracycline resistance operon to effectively repress and derepress transcription from one of the strongest mammalian promoter sequences known (1,2).

Specific Activation
The T-REx™ System uses a repressor mechanism that blocks transcription from the powerful CMV promoter in the absence of tetracycline. Because the T-REx™ System elements do not use viral transactivators, you can achieve high-level expression from the complete CMV promoter without secondary, non-specific activation of host genes.

The T-REx™ Mechanism
The T-REx™ transcriptional control elements are illustrated in Figure 1. Two tetracycline operator sequences (TetO2) have been inserted between the TATA box of the CMV promoter and the transcriptional start site. The TetO2 sequence itself has no effect on expression. When the tetracycline repressor protein (TR) is present, it effectively binds the TetO2 sites and blocks transcription initiation. Tetracycline added to the culture medium binds to, and changes the conformation of, the TR protein. This change causes the TR protein to release the TetO2 sites, derepressing transcription from the CMV promoter. The result is high-level expression of the gene of interest (Figure 2). Expression levels can be modulated based on the tetracycline concentration and can be induced to levels that are achieved with constitutive CMV expression vectors.
T-REx™ is a powerful inducible mammalian expression system that allows you to regulate expression from the complete human cytomegalovirus (CMV) enhancer-promoter. T-REx™inducible expression vectors offer the following features:

• Complete CMV enhancer-promoter sequence containing two copies of the tetracycline operator TetO2 sequence for high-level regulated expression
• Zeocin™ or hygromycin resistance gene for effective selection of stable mammalian cell lines
• Large multiple cloning site to simplify cloning

In addition, pcDNA™4/TO/myc-His offers a c-myc epitope for rapid detection of the recombinant protein with an Anti-myc Antibody and a polyhistidine (6xHis) sequence for simple purification of the recombinant protein with nickel-chelating resin and detection with Anti- His(C-term) Antibody.

The regulatory vector, pcDNA™6/TR, is provided for high-level expression of the tetracycline repressor (TR) protein. This vector expresses the Blasticidin resistance gene for rapid selection of mammalian cell lines that stably express the TR protein.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Promoter: CMV⁄TO
Key Function: Regulated Expression
Protein Tag or Fusion: His Tag (6x), c-Myc Epitope Tag
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Zeocin™
Vector Type: pcDNA
Product Size: 1 kit
Inducing Agent: Tetracycline
Constitutive or Inducible System: Inducible

Contents & storage

The T-REx™ Core Kit includes 20 µg of pcDNA™4/TO or 20 µg each of pcDNA™4/TO/myc-His A, B, and C; a positive control vector, pcDNA™6/TR, and forward and reverse sequencing primers.

The T-REx™ Complete Kit includes the Core Kit plus 1 g Zeocin™ (100 mg/ml), 50 mg of Blasticidin, and 5 g of tetracycline.

All vectors are supercoiled and provided lyophilized.

Store all components at -20°C. Zeocin™ and tetracycline should be protected from exposure to light. All components are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols