Invitrogen™

β-Gal Assay Kit

Catalog number:  K145501

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The β-Gal Assay Kit provides the reagents required to quickly measure the levels of active β-galactosidase expressed in cells transfected with plasmids expressing the lacZ gene. lacZ is a bacterial gene often used as a reporter construct in eukaryotic transfection experiments. The gene product of lacZ, β-galactosidase, is resistant to proteolysis in cellular lysates and its activity is easily assayed. β-galactosidase catalyzes the hydrolysis of β-galactosidase such as ortho-nitrophenyl-D-galactopyranoside (ONPG). Hydrolysis of ONPG to the ONP anion produces a bright yellow color with a peak absorbance at 420 nm that can be quantified using a spectrophotometer.
10.7 gene expression
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: β-Gal (lacZ) Assay
Detection Method: Colorimetric
Product Size: 80 mL
Substrate: ONPG
Substrate Properties: Chemical Substrate
Substrate Type: Beta-Gal Substrate
Target Enzyme: Beta-Galactosidase

Contents & storage

The β-Gal Assay Kit contains 10X PBS (phosphate buffered saline), lysis buffer, cleavage buffer, β-mercaptoethanol, ONPG, stop buffer. Store ONPG at -20°C. All other components are stored at either room temperature or +4°C. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products