During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

Tango™ DRD1-bla U2OS Cells

Catalog number: K1485

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Tango™ DRD1-bla U2OS cells contain the human Dopamine Receptor 1 (DRD1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The Tango™ DRD1-bla U2OS cell line has been functionally validated for Z'-factor and EC50 concentrations of dihydrexidine. The DRD1 cells have been tested for assay performance under variable assay conditions. These data are available in the Validation & Assay Performance Summary in the Manuals and Brochures section.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: DRD1
Accession Number: NM_000794.2
Cell Line: U2OS
Cell State: Dividing Cells
Cytoplasmic Tail: Tail from AVPR2 GPCR
Detection Method: Fluorescent
Druggable Target: GPCRs
Population Status: Single Cell Clone Origin
Product Line: Tango™
Product Size: 2 x 106 cells
Reporter: BLA (Beta-Lactamase)
System: Tango™

Contents & storage

(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
o Tango™ DRD1-bla U2OS cells
o Tango™ DRD1-bla U2OS Assay Protocol
o Certificate of Analysis
The Tango™ DRD1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.

Documents

Manuals & protocols