During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.
CellSensor™ NFAT-bla RA-1 Cell Line
Catalog number:  K1490
 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
B cell receptor (BCR)-mediated signaling pathways are important for B cell proliferation and differentiation. Abnormal B cell signaling has been linked to various diseases such as lupus, lymphoma, and various immune disorders. Antigen binding to the BCR promotes the activation of several protein tyrosine kinases (PTK), which leads to phosphorylation of the BCR complex and recruitment and activation of the PTK Syk. This in turn promotes phosphorylation of PLCγ, Shc, and Vav. Additionally, the Tec family member Btk is recruited to the plasma membrane, where it is involved in activation of PLCγ. Initiation of B lymphocyte activation is dependent on the tyrosine phosphorylation-dependent formation of multimolecular effector protein complexes that activate downstream signaling pathways including PKC/Ca2+/NFAT. This cell-based assay has been thoroughly tested and validated by Invitrogen and is suitable for immediate use in screening applications. The following information illustrates the comprehensive assay testing completed and validation of assay performance under optimized conditions (Figure 1).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell Line: RA-1
Cell State: Dividing Cells
Detection Method: Fluorescent
Druggable Target: Signaling Pathway
Product Line: CellSensor™
Product Size: 1 vial
System: CellSensor™
Contents & storage
CellSensor® NFAT-bla RA-1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Documents
Manuals & protocols
Recommended products