Related applications: RNA Extraction
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Sample Type (General): | Blood |
---|---|
High Throughput Compatibility: | Not High Throughput-Compatible (Manual) |
Downstream Application: | Microarray Analysis, Northern Blotting, Nuclease Protection Assays, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction |
Final Product: | Total RNA |
Format: | Spin Column |
Isolation Technology: | Silica Spin Column |
Number of Reactions: | 50 Preps |
Product Line: | PureLink™ |
Product Size: | 50 preps |
Purification Time: | 20 min |
Quantity: | 50 |
Scale: | Mini |
Starting Material (Amount): | 50 to 500 µl blood, Up to 3.5 x 10^6 leukocytes |
System: | PureLink™ |
Yield (up to): | 1000 µg |
Shipping Condition: | Room Temperature |
Catalog number: | Q10213 |
Size | 1,000 assays |
Price | USD 384.00 |
Add to favorites | |
Catalog number: | 11754050 |
Size | 50 reactions |
Price | USD 536.00 |
Add to favorites | |
Catalog number: | AM9780 |
Size | 250 mL |
Price | USD 73.00 |
Add to favorites | |