During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

LanthaScreen® STAT5 (JAK2 V617F) U2OS Cell Line

Catalog number: K1588

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

LanthaScreen® GFP Cellular Assays allow for the analysis of post-translational modifications for a number of target proteins in an live-cell context. The JAK2⁄STAT5 signaling pathway plays an essential role in blood cell formation in response to cytokines such as GM-CSF, IL-3, and EPO. The recent discovery of an activating mutation in JAK2 (V617F) present in high percentage of myeloproliferative disease (MPD) patients suggests that this mutant JAK2 activity is a potential therapeutic target for certain forms of MPD. The assay described in this summary makes use of a cell line engineered for expression of the constitutively-active mutant kinase JAK2 V617F. By co-expressing GFP-STAT5 in this background, the phosphorylation state of STAT5 (specifically residue Tyr 694⁄699) can be modulated with JAK2 V617F inhibitors and analyzed in cell lysates using an anti-STAT5 [pTyr 694⁄699] and LanthaScreenTM terbium-anti-mouse antibody pair. GFP-STAT5 Lentivirus was transduced into U2OS cells followed by selection with Blasticidin. The selected pool was then transfected with a GST-JAK2 V617F construct using LipofectamineTM LTX, followed by selection with Geneticin. This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as sorting marker, and has been screened for the constitutive expression of GFP-STAT5 and GST-JAK2 V617F. Using a lytic TR-FRET immuno-assay, this cell line is validated for IC50 and Z' under optimized conditions using JAK Inhibitor I (Pyridone 6) as a small molecule inhibitor for JAK2 V617F-mediated GFP-STAT5 phosphorylation. This assay has also been tested for assay performance under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance and lysis⁄equilibration time.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: STAT5A
Alternate Agonists: Constitutive
Antagonists: JAK Inhibitor
Cell Line: U2OS
Cell State: Dividing Cells
Detection Method: Fluorescent
Druggable Target: Kinases, Signaling Pathway
Optimal Response: Constitutively Active
Primary Agonist: Constitutive
Product Line: LanthaScreen®
Product Size: 1 vial
Readout: TR-FRET
Reporter: GFP (EmGFP)
System: LanthaScreen®

Contents & storage

LanthaScreen® STAT5 (JAK2 V617F) U2OS cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.

Documents

Manuals & protocols