During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.
CellSensor™ HRE-bla ME-180 Cell Line
Catalog number:  K1644
 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The CellSensor® HRE-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the hypoxia response element (HRE) stably integrated into ME-180 cells. To construct this cell line, the HRE-bla construct was transduced into ME-180 cells by lentivirus. Flow cytometry was used to isolate cells responsive to deferoxamine. This cell line is validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z' and EC50 concentrations for cobalt chloride and deferoxamine. The CellSensor® HRE-bla ME-180 cell line is responsive to deferoxamine and cobalt chloride and can be used to probe the hypoxia signaling pathway. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell State: Dividing Cells
Druggable Target: Signaling Pathway
Product Line: CellSensor™
Product Size: 1 vial
System: CellSensor™
Contents & storage
CellSensor® Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Documents
Manuals & protocols