During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

CellSensor™ ISRE-bla Jurkat Cell Line

Catalog number:  K1656

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CellSensor® ISRE-bla Jurkat cell line contains a beta-lactamase reporter gene under control of the interferon-stimulated response element (ISRE) stably integrated into Jurkat cells. To construct this cell line, the ISRE-bla construct was transduced into Jurkat cells by lentivirus. Flow cytometry was used to isolate cells responsive to IFNα. This cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as ' and EC50 concentrations of IFN&alpha. The CellSensor® ISRE-bla Jurkat cell line is responsive to Interferon alpha (IFN-alpha) and Interferon beta and can be used to probe the type I interferon-induced JAK-STAT signaling pathways. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can also be tested for dose response against this cell line. ISRE-bla Jurkat cells were treated with agonist IFN-alpha; over the indicated concentration range in a 384-well format. Cells were incubated for 5 hours with agonist and 0.5% DMSO and then combined with LiveBLAzer™-FRET B/G Substrate (CCF4-AM) for 2 hours. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader, and the 460/530 ratios were plotted against the concentration of the agonist. Academic and non-profit customers, please inquire for special pricing.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell State: Dividing Cells
Druggable Target: Cytokines, Signaling Pathway
Product Line: CellSensor™
Product Size: 1 vial
System: CellSensor™

Contents & storage

CellSensor® Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.

Documents

Manuals & protocols